β-Sitosterol ≥70%, crystalline solid
Dodavatel: Cayman Chemical
24α-ethyl Cholesterol is a naturally occurring plant sterol with antioxidant, anticancer, anti-inflammatory, angiogenic, chemopreventive, and immunomodulatory activities.
It inhibits the absorption of dietary and endogenously-produced cholesterol from the small intestine, reducing blood cholesterol concentrations. Because of its nutraceutical benefits, 24α-ethyl cholesterol has been used as a food additive intended to lower LDL cholesterol. 24α-ethyl Cholesterol (0,1 to 100 μM) can dose-dependently induce adipogenesis and lipolysis in rat primary preadipocytes as well as stimulate glucose uptake in differentiated adipocytes.
Synonyms: Azuprostat, Betaprost, Cupreol, NSC 18173, NSC 49083, NSC 8096, Rhamnol, SKF 14463, 22,23-dihydro-Stigmasterol.
Vzorec:
C₂₉H₅₀O |
MDL Number:
MFCD00003631 CAS číslo: 83-46-5 |
Výsledky specifikačního testu
Purity | ≥95% |
Form | Crystalline Solid |
Storage | Store at −20 °C |
Stability | As supplied, 2 years from the QC date provided on the Certificate of Analysis, when stored properly |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...