Tisk…

N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride, white powder

Dodavatel: MP Biomedicals

Synonyms: Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, Bz-Arg-pNA·HCl, Bz-Arg-PNA.HCl, N-α-Benzoyl-L-Arginine p-Nitroanilide Hydrochloride, (S)-N-(5-Guanidino-1-((4-nitrophenyl)amino)-1-oxopentan-2-yl)benzamide hydrochloride

0215482980 0215482925 0215482983
ICNA0215482980EA 5800 CZK
ICNA0215482980 ICNA0215482925 ICNA0215482983
N-α-Benzoyl-L-arginine 4-nitroanilide hydrochloride, white powder
N-alpha-Benzoyl-L-arginine 4-nitroanilide hydrochloride

Chromogenic substrate for trypsin and other proteolytic enzymes.


Soluble in methanol (50 mg/ml-clear, colorless solution).


Upozornení: Research Use Only (RUO).

Vzorec: C₁₉H₂₃ClN₆O₄
MW: 434,881 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00063682
CAS číslo: 21653-40-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR