Tisk…
Paraformaldehyd 16% (w/v) in aqueous solution methanol-free
Dodavatel: MP Biomedicals
Synonyms: Polyoxymethylene
Packaged under nitrogen.
0219998320 0219998380
ICNA0219998320EA 2240 CZK
ICNA0219998320 ICNA0219998380
Paraformaldehyd 16% (w/v) in aqueous solution methanol-free
Paraformaldehyd
Methanol free Paraformaldehyde is ready to use solution used as a fixative for electron microscopy.
Recommended for use in the preparation of formalin fixatives for tissues or cells when the samples are to be used in florescence studies.
Vzorec: (CH₂O)n
Bod tání: 123
Bod vzplanutí: 70
Storage Temperature: Refrigerator
MDL Number: MFCD00133991
CAS číslo: 30525-89-4
UN: 2213
ADR: 4.1 III
Merck Index: 13,07096