Tisk…
Ecolume™ liquid scintillation cocktail, MP Biomedicals
Dodavatel: MP Biomedicals
0188247001 0188247004
ICNA0188247001EA 9290 CZK
ICNA0188247001 ICNA0188247004
Ecolume™ liquid scintillation cocktail, MP Biomedicals
Scintilační koktejly
EcoLume™ is a non-toxic, non-hazardous, non-flammable liquid scintillation counting solution. Using EcoLume™ eliminates other toxic cocktails from your laboratory. At the same time, you make a commitment to reduce waste from our environment.
  • Environmentally safe
  • Non-toxic and non-flammable