Tisk…

Spermidine free base ∼99%, waxy solid

Dodavatel: MP Biomedicals

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

0215206801 0215206805 0215206825
ICNA0215206801EA 1180 CZK
ICNA0215206801 ICNA0215206805 ICNA0215206825
Spermidine free base ∼99%, waxy solid
Spermidine

Spermidine binds and precipitates DNA; may be used for purification of DNA binding proteins. Stimulates T4 polynucleotide kinase activity. It has a role in cell growth processes. It can be used in electroporation while transferring the DNA into the cell under the electrical impulse.


It is also plant growth regulator, promoting somatic embryogenesis.


Upozornení: Research Use Only (RUO).

Vzorec: C₇H₁₉N₃
MW: 145,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00008229
CAS číslo: 124-20-9

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard
Purity ~99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR