Tisk…

Cresyl violet (acetate), dark green powder

Dodavatel: MP Biomedicals

Synonyms: Cresyl echt violet, Cresyl violet, Cresyl fast violet, 5,9-Diaminobenzo[a]phenoxazin-7-ium acetate, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt, Cresyl Violet acetate, Gentian violet

0215072701 0215072705
ICNA0215072701EA 766 CZK
ICNA0215072701 ICNA0215072705
Cresyl violet (acetate), dark green powder
Cresyl violet (acetate)

Cresyl violet acetate is used for staining neve tissue and nerve cells and fresh tumor tissue for biopsy.

Vzorec: C₁₈H₁₅N₃O₃
MW: 321.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013151
CAS číslo: 10510-54-0

Objednat

Výsledky specifikačního testu

Identity Test Passes
λmax (ethanol) 596 - 602 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR