Sulfid sodný nonahydrát 98.0-103.0%, tan powder ACS
Dodavatel: MP Biomedicals
Synonyms:
Sodium sulphide nonahydrate
0215258090
0215258080
ICNA0215258090EA
2150
CZK
ICNA0215258090
ICNA0215258080
Sulfid sodný nonahydrát 98.0-103.0%, tan powder ACS
Sulfid sodný nonahydrát
Sodium sulfide is used in the formation of surface functionalized cadmium sulfide quantum dots. Also employed in the pulp and paper industry.
Soluble in water; slightly soluble in alcohol; insoluble in ether.
Vzorec:
Na₂S·9H₂O MW: 240,18 g/mol |
MDL Number:
MFCD00149184 CAS číslo: 1313-84-4 UN: 1849 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...