Tisk…

Cyclophosphamide monohydrate ≥98%, white crystalline powder

Dodavatel: MP Biomedicals
Nebezpečí

Synonyms: 2-(Bis(2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide hydrate, Cyclophosphamide hydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-1,3,2-oxazaphosphorin-2-oxide

0215074901 0215074905 0215074910 0215074925
ICNA0215074901EA 2300 CZK
ICNA0215074901 ICNA0215074905 ICNA0215074910 ICNA0215074925
Cyclophosphamide monohydrate ≥98%, white crystalline powder
Cyclophosphamide monohydrate

Cyclophosphamide monohydrate used for induction chemotherapy to study predictive and prognostic values of tumor MGMT gene expression, to study its effects on apoptosis and cell cycle progression and its cardiotoxicity during pretransplant conditioning in blood stem cell transplantation has been reported.


Soluble in water (40 mg/ml - clear, colorless solution). Slightly soluble in ethanol, benzene, ethylene glycol, carbon tetrachloride, dioxane; sparingly soluble in ether and acetone.

Vzorec: C₇H₁₇Cl₂N₂O₃P
Storage Temperature: Refrigerator
MDL Number: MFCD00149395
CAS číslo: 6055-19-2

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 44 - 54 °C
pH (1% aq soln) 4 - 6


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR