Chlorid lantanitý heptahydrát, clear to white crystalline powder ACS
Dodavatel: MP Biomedicals
|
Varování
|
0215253480
0215253425
0215253490
ICNA0215253480EA
4110
CZK
ICNA0215253480
ICNA0215253425
ICNA0215253490
Chlorid lantanitý heptahydrát, clear to white crystalline powder ACS
Chlorid lantanitý heptahydrát
Calcium channel blocker.
- Calcium channel blocker
Lanthanum chloride is used to precipitate phosphate from solutions. Lanthanum chloride is also used in biochemical research to block the activity of divalent cation channels, mainly calcium channels.
Vzorec:
LaCl₃·7H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149756 CAS číslo: 10025-84-0 |
Výsledky specifikačního testu
Assay | 64.5 - 70.0% LaCl3 |
Insoluble Matter | ≤0.01% |
Calcium (Ca) | ≤0.001% |
Magnesium (Mg) | ≤10ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...