Fast blue B salt zinc double salt, powder C.I. 37235
Dodavatel: MP Biomedicals
0215020110
0215020125
0215020180
ICNA0215020110EA
2470
CZK
ICNA0215020110
ICNA0215020125
ICNA0215020180
Fast blue B salt zinc double salt, powder C.I. 37235
Fast blue B salt zinc double salt
Fast blue B salt is useful for TLC of Aflatoxins.
Soluble in water( 4 mg/4 ml - clear solution).
Upozornení: Research Use Only (RUO).
Vzorec:
C₁₄H₁₂Cl₄N₄O₂Zn |
MDL Number:
MFCD00075203 CAS číslo: 14263-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...