Tisk…

8-Anilino-1-naphthalenesulphonic acid, dark powder

Dodavatel: MP Biomedicals
Varování

Synonyms: 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid

0210563301 0210563305 0210563310 0210563325
ICNA0210563301EA 1140 CZK
ICNA0210563301 ICNA0210563305 ICNA0210563310 ICNA0210563325
8-Anilino-1-naphthalenesulphonic acid, dark powder
8-Anilino-1-naphthalenesulphonic acid

8-Anilino-1-naphthalene Sulphonic Acid is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins.


ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants. It is a hydrophobic fluorescent probe for undenatured proteins on polyacrylamide gels.


Soluble in DMF (50 mg/ml). Soluble in 1 N NaOH, methanol (50 mg/ml - clear brownish green solution), chloroform (30 mg/ml in a 1:1 mixture produces a clear olive green solution); slightly water soluble.


Upozornení: Research Use Only (RUO).

Vzorec: C₁₆H₁₃NO₃S
MW: 299,35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003998
CAS číslo: 82-76-8

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR