Caffeic acid ≥98%, yellow powder
Dodavatel: MP Biomedicals
Synonyms:
3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
Caffeic acid is used as an anti-oxidant.
Inhibits the synthesis of leukotrienes that are involved in immunoregulation, inflammation and allergy. Inhibits Cu2+-induced LDL oxidation.
Soluble in ethanol (50 mg/ml) (Lit.).
Vzorec:
C₉H₈O₄ MW: 180,16 g/mol |
MDL Number:
MFCD00004392 CAS číslo: 331-39-5 |
Výsledky specifikačního testu
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...