Vitamin A palmitate in corn oil, >1,000,000 IU/g, clear yellow liquid
Dodavatel: MP Biomedicals
|
Nebezpečí
|
Synonyms:
Retinyl palmitate, Retinol palmitate, all-trans-Retinyl palmitate
0210326325
0210326380
ICNA0210326325EA
2850
CZK
ICNA0210326325
ICNA0210326380
Vitamin A palmitate in corn oil, >1,000,000 IU/g, clear yellow liquid
Vitamin A palmitate
- Presentation : Clear Yellow Liquid
Storage: under nitrogen.
Vzorec:
C₃₆H₆₀O₂ MW: 524,87 g/mol Bod tání: 28,..29 °C (lit.) Bod vzplanutí: 194 °C (381,2 °F) (lit.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00019414 CAS číslo: 79-81-2 |
Výsledky specifikačního testu
Identity Test | Passes |
Activity | >1,000,000 IU/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...