Vitamin A acetate, ~500,000 IU/gm, yellow powder
Dodavatel: MP Biomedicals
|
Varování
|
Synonyms:
all-trans-Retinyl acetate, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl acetate
The product contains vitamin A acetate (approximately 21%) finely dispersed in a starch-coated matrix of gelatin (approximately 60%), sucrose (approximately 14%) and corn starch (approximately 5%). BHT and BHA are added as antioxidants.
- Concentration: Vitamin A Content: >500,000 IU/g
- Specific Activity : ~500,000 IU/gm
- Presentation : Yellow Powder
- Particle Size : 100% through sieve No. 2; minimum 90% through sieve No. 40; Maximum 15% through sieve No. 100.
- Loss on Drying : ≤8%
The product contains vitamin A acetate (approximately 21%) finely dispersed in a starch-coated matrix of gelatin (approximately 60%), sucrose (approximately 14%) and corn starch (approximately 5%). BHT and BHA are added as antioxidants.
Vitamin A acetate induces cell differentiation and inhibits cell proliferation.
Vzorec:
C₂₂H₃₂O₂ MW: 328,49 g/mol Bod tání: 57,..58 °C (lit.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00019413 CAS číslo: 127-47-9 |
Výsledky specifikačního testu
Identity Test | Passes |
Loss on Drying | <8% |
Vitamin A Content | >500,000 IU/g |
Microbiological Purity | Satisfactory |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...