Tisk…

Histamine diphosphate, crystalline powder

Dodavatel: MP Biomedicals
Nebezpečí

Synonyms: 2-(4-Imidazolyl)ethylamine diphosphate salt, 2-(1H-Imidazol-4-yl)ethan-1-amine bis(phosphate), Histamine phosphate

0210034301 0210034305 0210034325
ICNA0210034301EA 1220 CZK
ICNA0210034301 ICNA0210034305 ICNA0210034325
Histamine diphosphate, crystalline powder
Histamine diphosphate

Histamine Diphosphate Monohydrate is potent vasodilater.


Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilizes Ca2+; H2â‚‚ activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.


Endogenous H1 and H2 histamine receptor agonist; H1 activation mobilises Ca2+; H22‚ activation stimulates adenylate cyclase activity in neurons; activates nitric oxide synthetase; potent vasodilator.


Soluble in water (50 mg/mL - clear, colorless to almost colorless solution).


Upozornení: Research Use Only (RUO).

Vzorec: C₅H₉N₃·2H₃PO₄
MW: 307.14 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00064316
CAS číslo: 51-74-1

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard
Loss on Drying Report result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR