Tisk…

Carboxymethoxylamine hemihydrochloride, white crystalline powder

Dodavatel: MP Biomedicals
Varování

02150104.2 0215010401 0215010405
ICNA02150104.2EA 1250 CZK
ICNA02150104.2 ICNA0215010401 ICNA0215010405
Carboxymethoxylamine hemihydrochloride, white crystalline powder
Carboxymethoxylamine hemihydrochloride

Soluble in 100 mg/ml of water.


(Aminooxy)acetic Acid Hemihydrochloride undergoes condensation with aldehydes and ketones to form oximes. It inhibits aminobutyrate aminotransferase activity in vivo, thereby raising the level of gamma-aminobutyric acid in tissues.


Soluble in 100 mg/ml of water.


Upozornení: Research Use Only (RUO).

Vzorec: C₄H₁₁ClN₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00012955
CAS číslo: 2921-14-4

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard
Melting Point Report Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR