Tisk…

D-α-Phenylalanine ≥99%, white powder

Dodavatel: MP Biomedicals

0210261605 0210261625 0210261680
ICNA0210261605EA 1350 CZK
ICNA0210261605 ICNA0210261625 ICNA0210261680
D-α-Phenylalanine ≥99%, white powder
D-α-Phenylalanine

Soluble in water (30 mg/ml). Also soluble in 1 M HCl (50 mg/ml - clear to slightly hazy solution).


Soluble in water (30 mg/ml). Also soluble in 1 M HCl (50 mg/ml - clear to slightly hazy solution).


Upozornení: Research Use Only (RUO).

Vzorec: C₉H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00004270
CAS číslo: 673-06-3

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Optical Rotation (c=2, H2O) +35° ± 5°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR