DL-Lithium lactate ≥98%, off-white powder
Dodavatel: MP Biomedicals
Synonyms:
DL-Lactic acid lithium salt, (±)-2-Hydroxypropanoic acid lithium salt, Lithium 2-hydroxypropanoate, (±)-2-Hydroxypropionic acid lithium salt, (±)-Lactic acid lithium salt
L-(+)-Lactic acid lithium salt is a fermentation acid from milk or carbohydrates.
Soluble in water.
Upozornení: Research Use Only (RUO).
Vzorec:
H₃CCH(OH)COOLi MW: 96,01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065399 CAS číslo: 867-55-0 |
Výsledky specifikačního testu
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...