Tisk…

Indol-3-acetic acid ~99, off-white powder

Dodavatel: MP Biomedicals

Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid

0210203705 0210203725 0210203780
ICNA0210203705EA 733 CZK
ICNA0210203705 ICNA0210203725 ICNA0210203780
Indol-3-acetic acid ~99, off-white powder
Indol-3-acetic acid

Indole-3-acetic acid (IAA) is the most common, naturally-occurring, plant growth regulator - as a principal auxin of higher plants. It is the best known of the auxins, and has been the subject of extensive studies by plant physiologists. Chemically, IAA is a carboxylic acid in which the carboxyl group is attached through a methylene group to the C-3 position of an indole ring.


Indole-3-acetic acid, one of its derivatives bis-2-indolyl disulphide has been used to synthesize tyrosin kinase inhibitors. It can also be used as a starting material to synthesize DBI receptor ligands. Methyl ester of indole-3-acetic acid reacts with dimethylamine and the resulting amide on reduction with Lithium aluminium hydride gives the hallucinogen N,N-Dimethyltryptamine. Used as a plant growth regulator as a principle auxin of higher plants. Can also be used as a starting material to synthesize DBI receptor ligands.


Soluble in methanol, ethanol (5%), Sparingly soluble in water, chloroform. Soluble in acetone, ether and freely soluble in alcohol.

Vzorec: C₁₀H₉NO₂
MW: 175,19 g/mol
MDL Number: MFCD00005636
CAS číslo: 87-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR