Tisk…
Guanosine-5′-monophosphate ≥98%, white powder
Dodavatel: MP Biomedicals
Synonyms: Guanosine 5'-Monophosphate hydrate, GMP, 5'-Guanylic acid, Guanosine 5'-monophosphoric acid
0210190880 0210190890 0210190891
ICNA0210190880EA 929 CZK
ICNA0210190880 ICNA0210190890 ICNA0210190891
Guanosine-5′-monophosphate ≥98%, white powder
Guanosine-5′-monophosphate
Guanosine-5'-monophosphate is an ester of phosphoric acid with the nucleoside guanosine.
Guanosine-5′-monophosphate (GMP) has be used to study modulation of glutamatergic neurotransmission.
Soluble in water (5% clear solution)
Upozornení: Research Use Only (RUO).
Vzorec: C₁₀H₁₄N₅O₈P
MW: 363,20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00057066
CAS číslo: 85-32-5
Objednat
Výsledky specifikačního testu
Identity Test Passes
Purity ≥98%
FTIR Conforms to Reference