Tisk…

D-(+)-Glucosamine hydrochloride, white powder

Dodavatel: MP Biomedicals

0210178225 0210178280 0210178290
ICNA0210178225EA 1510 CZK
ICNA0210178225 ICNA0210178280 ICNA0210178290
D-(+)-Glucosamine hydrochloride, white powder
D-(+)-Glucosamine hydrochloride

Supplementation of cell culture media with glucosamine hydrochloride may enhance N-glycosylation of secreted proteins and affect differentiation of cell lines such as chondocytes and stem cells.


Soluble in water (100 mg/ml).


Upozornení: Research Use Only (RUO).

Vzorec: C₆H₁₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD00135831
CAS číslo: 66-84-2

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard
+72° ± 10° (c=1, water) ≤0,5%
Loss on Drying Optical Rotation
Residue on Ignition ≤0.1%
Arsenic ≤0.5 ppm
Heavy Metals ≤10 ppm
Iron ≤20 ppm
Solubility (5% aq soln) Clear and Colourless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR