Tisk…

DL-Ethionine ∼99%, white powder

Dodavatel: MP Biomedicals

Synonyms: 2-Amino-4-(ethylthio)butanoic acid

0210166201 0210166205
ICNA0210166201EA 2600 CZK
ICNA0210166201 ICNA0210166205
DL-Ethionine ∼99%, white powder
DL-Ethionine

DL-Ethionine is a racemic mixture of the anti-methylation agent L-ethionine and its enantiomer D-ethionine. DL-Ethionine is used as a dietary supplement to accelerate cholangiocarcinogenesis in vivo. DL-Ethionine is used to induce oxidative stress in liver to study the levels and activities of anti-oxidative enzymes and compounds such as glutathione.


Soluble in 1M hydrochloric acid (50 mg/ml - very slightly hazy colorless solution).


Upozornení: Research Use Only (RUO).

Vzorec: C₆H₁₃NO₂S
MW: 163,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063102
CAS číslo: 67-21-0

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR