Tisk…

Maltotriose 98%

Dodavatel: Thermo Fisher Scientific

Synonyms: Amylotriose, α-1,4-Glucotriose, α-D-Glc-(1→4)-α-D-Glc-(1→4)-D-Glc, (2R,3R,4R,5R)-4-(((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal

J66491.03 J66491.06
J66491.03EA 3220 CZK
J66491.03 J66491.06
Maltotriose 98%
Maltotriose

Maltotriose is a matrose regulon inducer in Escherichia coli.


Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.


Upozornení: Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Vzorec: C₁₈H₃₂O₁₆
Bod tání: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00006629
CAS číslo: 1109-28-0
EINECS: 214-174-2

Objednat

Výsledky specifikačního testu

Synonyms Alpha-1,4-Glucotriose
Form Powder
Solubility Soluble in Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR