Tisk…

Kyselina stearová ≥99%, white crystalline powder

Dodavatel: MP Biomedicals

Synonyms: Octadecanoic acid, Octadecylic acid

0210111705 0210111710 0210111725 0210111780
ICNA0210111705EA 1130 CZK
ICNA0210111705 ICNA0210111710 ICNA0210111725 ICNA0210111780
Kyselina stearová ≥99%, white crystalline powder
Kyselina stearová

Stearic acid, an anti-inflammatory agent, is the hydrogenation product of linoleic acid.


Stearic acid has been shown to induce a decrease of cell growth and adhesion and increase hydrophobic properties of collagen films. Stearic Acid has been noted to induce an increase in ICAM-1 expression in cells, via an increase in phosphorylation of the transcriptional regulator NFκB.


Soluble in alcohol, benzene, chloroform, acetone, carbon tetrachloride, carbondisulphide; very slightly soluble in water.

Vzorec: C₁₈H₃₆O₂
MDL Number: MFCD00002752
CAS číslo: 57-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR