Tisk…

3-Amino-1H-1,2,4-triazole, white powder

Dodavatel: MP Biomedicals
Nebezpečí

0210064710 0210064780
ICNA0210064710EA 953 CZK
ICNA0210064710 ICNA0210064780
3-Amino-1H-1,2,4-triazole, white powder
3-Amino-1H-1,2,4-triazole

3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function.


3-Amino-1,2,4-triazole is used as a reagent for the detection of amino acid oxidase. Also a reagent for the determination of tryptophan in proteins. Commercial grade 3-amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a herbicide and cotton defoliant.


Soluble in water (50 mg/ml), methanol, ethanol, and chloroform. Sparingly soluble in ethyl acetate. Insoluble in ether or acetone.

Vzorec: C₂H₄N₄
Storage Temperature: Freezer
MDL Number: MFCD00053362
CAS číslo: 61-82-5

Objednat

Výsledky specifikačního testu

Identity Test Passes
FTIR Conforms to Standard
Melting Point 145 - 155 °C

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR