Tisk…

Aspergillus niger Catalase, MP Biomedicals

Dodavatel: MP Biomedicals

0210040250 0210040280
ICNA0210040250EA 3440 CZK
ICNA0210040250 ICNA0210040280
Aspergillus niger Catalase, MP Biomedicals
Enzymy

Catalase, fungal suspension from from Aspergillus niger is long-acting, extremely stable form of catalase that is active over a wide pH range. Composed of four protein subunits, each containing a heme group bound to its active site.


  • Fungal suspension
  • Caution: freezing of solutions is not recommended


Catalase is used for the removal of peroxides, the generation of oxygen and, in coupled systems, for the determination of metabolites e.g. uric acid. A very stable preparation particularly suitable for immobilisation.


One unit decomposes 1,0 µmole of hydrogen peroxide per minute at pH 7,0, 25 °C.

Objednat

Produktová specifikace

Název enzymu Catalase
Environmentally Preferable
Synonyma enzymů E.C. 1.11.1.6, H2O2:H2O2 oxidoreductase
Zdroj
Druhy Aspergillus niger
Čí. CAS 9001-05-2
Podmínky uskladnění 4 °C
Aktivita enzymu ≥1000 u/ml
Rozpustnost Miscible in water

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR