Tisk…

Hydrogenuhličitan sodný 99.7-100.3% (dried basis), white powder ACS

Dodavatel: MP Biomedicals

Synonyms: Sodium bicarbonate

02191435.5 0219143501
ICNA02191435.5EA 1230 CZK
ICNA02191435.5 ICNA0219143501
Hydrogenuhličitan sodný 99.7-100.3% (dried basis), white powder ACS
Hydrogenuhličitan sodný

Soluble in Water.

Vzorec: NaHCO₃
MW: 84.01
Bod tání: 270
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS číslo: 144-55-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Identity Test Passes
Assay 99,7 - 100,3% (dried basis)
Insoluble Matter ≤0,015%
Chloride (Cl) <0,003%
Phosphate (PO4) <0,001%
Sulphur Compounds (as SO4) <0,003%
Ammonium (NH4) <5 ppm
Magnesium (Mg) <0,005%
Potassium (K) <0,005%
Calcium (Ca) <0,02%
Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) <0,001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR