Polyethylene glycol, M.W. ∼ 3,400, white powder
Dodavatel: MP Biomedicals
Synonyms:
Poly(ethylene oxide), Macrogol, PEO, PEG, Carbowax
Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water. PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds. PEG is incompatible with phenol and may reduce the antimicrobial action of other preservatives.
- Auto Ignition: 581 °F (Lit.)
- Vapor Density: >1 (vs air) (Lit.)
- Vapor Pressure: <0.01 mmHg at 20 °C (Lit.)
PEG has been used in the precipitation of proteins, as a fusing agent in enhancing the effect of macrophages on hybridoma, in the separation and purification of biomolecules, and in induction of cell hybridization PEG has also been used to recover pathogenic viruses and human rotaviruses from water, oyster and sediment samples. PEG has demonstrated to have anticell proliferation and antitumor effects via induction of apoptosis.
Soluble in aqueous solution (100 mg/mL-clear, colorless solution)
Upozornení: Research Use Only (RUO).
Vzorec:
H(OCH₂CH₂)nOH Bod tání: 55…65 °C Hustota: 1,1…1.2 g/mL (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS číslo: 25322-68-3 |
Výsledky specifikačního testu
Identity Test | Passes |
FTIR | Conforms to Reference |
Melting Point | 55-65°C |
pH (5% aq soln) | 4.5-7.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...