6-Benzylaminopurine, powder
Dodavatel: MP Biomedicals
|
Varování
|
Synonyms:
6-BAP, 6-Benzyladenine, N6-Benzyladenine, Benzyl(purin-6-yl)amine
6-Benzylaminopurine is a plant cell division factor (a plant growth factor).
- E (270) = ~19,054 (methanol) (Lit.)
6-Benzylaminopurine, benzyl adenine (BAP) is a synthetic cytokinin which together with auxins elicits plant growth and development responses. BAP is a widely use cytokinin supplement to plant growth media such as Murashige and Skoog medium, Gamborg′s medium, and Chu′s N6 medium. Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables.
BAP is an inhibitor of respiratory kinase in plants, and increases post-harvest life of green vegetables.
E (270) = ~19,054 (methanol) (Lit.)
Soluble in glacial acetic acid (50 mg/ml). Soluble in concentrated ammonium hydroxide, 0,1 N Sodium hydroxide (10 mg/ml - slightly hazy and yellowish solution), methanol, or dioxane.
Upozornení: Research Use Only (RUO).
Vzorec:
C₁₂H₁₁N₅ MW: 225,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005572 CAS číslo: 1214-39-7 |
Výsledky specifikačního testu
Identity Test | Passes |
FTIR | Conforms to Standard |
Melting Point | 226 - 236 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...