Tisk…
HPLC columns, Polaris™ NH₂
Dodavatel: VARIAN
A2013020X020 A2013030X020 A2013050X020 A2013050X046 A2013100X020 A2013100X030 A2013100X046 A2013125X040 A2013150X020 A2013150X030 A2013150X040 A2013150X046 A2013250X020 A2013250X030 A2013250X046 A2013250X100 A2013250X212 A2014020X020 A2014030X020 A2014050X020 A2014050X030 A2014050X046 A2014100X020 A2014100X030 A2014100X046 A2014150X020 A2014150X030 A2014150X046 A2014250X020 A2014250X030 A2014250X046
VARIA2013020X020EA 17610 CZK
VARIA2013020X020 VARIA2013030X020 VARIA2013050X020 VARIA2013050X046 VARIA2013100X020 VARIA2013100X030 VARIA2013100X046 VARIA2013125X040 VARIA2013150X020 VARIA2013150X030 VARIA2013150X040 VARIA2013150X046 VARIA2013250X020 VARIA2013250X030 VARIA2013250X046 VARIA2013250X100 VARIA2013250X212 VARIA2014020X020 VARIA2014030X020 VARIA2014050X020 VARIA2014050X030 VARIA2014050X046 VARIA2014100X020 VARIA2014100X030 VARIA2014100X046 VARIA2014150X020 VARIA2014150X030 VARIA2014150X046 VARIA2014250X020 VARIA2014250X030 VARIA2014250X046
HPLC columns, Polaris™ NH₂
Kolony chromatografické
Polaris™ NH₂ is an aminopropylsilane-covered silica that can be used in either reversed or normal phase conditions. Exhibit higher efficiencies for carbohydrate analysis than ionic gel-based columns
  • Mobile phase is acetonitrile/water
  • Shorter gradient re-equilibration times
Monomolecular layer of aminopropylsilane chemically bonded to totally porous silica gel support, 10 µm in diameter.