Tisk…
CellMaxx™ Bovine transferrin (holo) (from Serum), MP Biomedicals
Dodavatel: MP Biomedicals
0215233580 0215233590
ICNA0215233580EA 3590 CZK
ICNA0215233580 ICNA0215233590
CellMaxx™ Bovine transferrin (holo) (from Serum), MP Biomedicals
Proteiny
CellMaxx™ Bovine Transferrin is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. Transferrin (HOLO) is an iron-rich product supplied as a heat-treated, lyophilised powder.
It is frequently used as a nutrient in cell and microbial culture and as a media supplement in bio-pharmaceutical production. Also, it can be used with a wide range of cell types.