Tisk…
Capillary columns, BP1 PONA
Dodavatel: Trajan Scientific and Medical
054818 054950
568-0237EA 40070 CZK
568-0237 568-0241
Capillary columns, BP1 PONA
Kolony chromatografické
These 100% Dimethyl Polysiloxane columns are designed for the analysis of petroleum products.
  • Non polar phase for PONA analysis
Suitable replacement for: Petrocol DH, DB-Petro, HP-PONA, AT-Petro, Elite-PONA, ZB-1, 007-1-100-0.5F, Rtx-1PONA, CP Sil PONA.
Max. temperature: 340 °C.