Tisk…

Sodium trifluoromethanesulphinate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Trifluoromethanesulphinic acid Sodium Salt, Langlois Reagent

454990010 454990050 454990250
ACRO454990010EA 931 CZK
ACRO454990010 ACRO454990050 ACRO454990250
Sodium trifluoromethanesulphinate 98%
Sodium trifluoromethanesulphinate
Vzorec: CHF₃O₂S
MW: 156,05 g/mol
Bod tání: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD03092989
CAS číslo: 2926-29-6

Objednat

Výsledky specifikačního testu

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR