Tisk…

Kyselina jodistá for analysis

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: ortho-Periodic acid, Orthoperiodic acid

453170250 453171000 453175000
ACRO453170250EA 1310 CZK
ACRO453170250 ACRO453171000 ACRO453175000
Kyselina jodistá for analysis
Kyselina jodistá
Vzorec: H₅IO₆
Bod tání: 122 °C
Hustota: 1,3875 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS číslo: 10450-60-9
UN: 3085
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39e1882cdf619581f467ea2429265a700b5205d5&vid=27e789fa0f30c08d62d5a8d0afc3acd8f6469c12&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder or crystals
Titration Iodimetric 99.0 to 101.0 %
Residue after ignition ≤0.05 %
Iodide (I) ≤10 ppm
Bromide and Bromate (as Cl) ≤200 ppm
Chloride and Chlorate (as Cl) ≤200 ppm
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤50 ppm
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm
Nickel (Ni) ≤50 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤100 ppm
Zinc (Zn) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm
Insoluble matter ≤0.01 %
Iodic acid ≤1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR