Tisk…

Chloromethylphosphonic acid 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

451600010 451600050
ACRO451600010EA 3420 CZK
ACRO451600010 ACRO451600050
Chloromethylphosphonic acid 98%
Chloromethylphosphonic acid
Vzorec: CH₄ClO₃P
Bod tání: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00013955
CAS číslo: 2565-58-4
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 88 to 92 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR