Tisk…

Diphenyliodonium hexafluorophosphate 97%

Dodavatel: Thermo Scientific
Nebezpečí

450910050 450910250 450911000
ACRO450910050EA 1770 CZK
ACRO450910050 ACRO450910250 ACRO450911000
Diphenyliodonium hexafluorophosphate 97%
Diphenyliodonium hexafluorophosphate
Vzorec: C₁₂H₁₀F₆IP
MW: 426,08 g/mol
Bod tání: 140…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00061398
CAS číslo: 58109-40-3
UN: 1759
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 138 to 143 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR