Tisk…

[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4'-Chloro-[1,1'-biphenyl]-4-sulphonyl chloride, 4'-Chlorobiphenyl-4-sulphonyl chloride

450330010 450330100
ACRO450330010EA 1350 CZK
ACRO450330010 ACRO450330100
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride 97%
[4-(4-Chlorophenyl)phenyl]sulphonyl chloride
Vzorec: C₁₂H₈Cl₂O₂S
MW: 287,17 g/mol
Bod tání: 109…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631918
CAS číslo: 20443-74-7
UN: 3261
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 106 to 112 °C
HPLC ≥96 % as diethylsulfonamide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR