Cytidine-5'-monophosphate (from yeast) 97%
Dodavatel: Thermo Fisher Scientific
449490010
449490050
ACRO449490010EA
2750
CZK
ACRO449490010
ACRO449490050
Cytidine-5'-monophosphate (from yeast) 97%
Cytidine-5'-monophosphate
Vzorec:
C₉H₁₄N₃O₈P Storage Temperature: Refrigerator |
MDL Number:
MFCD00006544 CAS číslo: 63-37-6 |
Výsledky specifikačního testu
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤3 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...