Tisk…

Benzylalkohol, meets analytical specification of Ph.Eur.

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

447000010 447000025
ACRO447000010EA 2460 CZK
ACRO447000010 ACRO447000025
Benzylalkohol, meets analytical specification of Ph.Eur.
Benzylalkohol
Vzorec: C₆H₅CH₂OH
MW: 108.14
Bod varu: 205 °C (760 mmHg)
Bod tání: –15 °C
Hustota: 1.044 g/cm³
Bod vzplanutí: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS číslo: 100-51-6
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Appearance Clear colorless Oily liquid
Infrared spectrum Conforms
Refractive index 1.5380 to 1.5410 (20°C, 589 nm)
Assay 98.0 to 100.5 %
Appearance of solution Clear colorless
Residue after evaporation ≤0.05 %
Related substances ≤0.3 % (Sum of other peaks with relative retention greater than that of benzylalcohol)
Acidity Passes test
Peroxide value ≤5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR