Tisk…

2',7'-Dichlorofluorescein ACS

Dodavatel: Thermo Fisher Scientific
Varování

446930010 446930100
ACRO446930010EA 1780 CZK
ACRO446930010 ACRO446930100
2',7'-Dichlorofluorescein ACS
2',7'-Dichlorofluorescein
Vzorec: C₂₀H₁₀Cl₂O₅
Hustota: 0,79 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005047
CAS číslo: 76-54-0

Objednat

Výsledky specifikačního testu

Appearance Light orange to red-brown Powder
Suitability (as adsorption indicator) Passes test
Clarity of alcohol solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR