Tisk…

2,3-Difluoroisonicotinic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 2,3-Difluoropyridine-4-carboxylic acid

446770010 446770050
ACRO446770010EA 2370 CZK
ACRO446770010 ACRO446770050
2,3-Difluoroisonicotinic acid 97%
2,3-Difluoroisonicotinic acid
Vzorec: C₆H₃F₂NO₂
Bod tání: 165…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09031174
CAS číslo: 851386-31-7

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
NMR (1H-NMR) Consistent with structure
Titration with NaOH ≥96.0 to ≤104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR