1-Boc-3-fluoro-4-piperidone 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Piperidinecarboxylic acid, 3-fluoro-4-oxo-, 1,1-dimethylethyl ester, tert-Butyl 3-Fluoro-4-oxo-1-piperidinecarboxylate, tert-Butyl 3-fluoro-4-oxopiperidine-1-carboxylate, 3-Fluoro-4-oxopiperidine-1-carboxylic acid tert-butyl ester, 1-(tert-Butoxycarbonyl)-3-fluoro-4-piperidone, 3-Fluoro-4-oxo-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-fluoro-4-piperidone
446510010
446510050
ACRO446510010EA
6950
CZK
ACRO446510010
ACRO446510050
1-Boc-3-fluoro-4-piperidone 97%
1-Boc-3-fluoro-4-piperidone
Vzorec:
C₁₀H₁₆FNO₃ MW: 217,24 g/mol Bod tání: 75…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD10566554 CAS číslo: 211108-50-8 |
Výsledky specifikačního testu
Appearance | White to cream to light brown Powder or solid |
Infrared spectrum | Conforms |
Melting point | 73 to 78 °C |
Water | ≤2.5 % |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...