4-Formylbenzeneboronic acid pinacol ester 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
445040050
445040250
ACRO445040050EA
2560
CZK
ACRO445040050
ACRO445040250
4-Formylbenzeneboronic acid pinacol ester 97%
4-Formylbenzeneboronic acid pinacol ester
Vzorec:
C₁₃H₁₇BO₃ MW: 232.086 g/mol Bod tání: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD04972375 CAS číslo: 128376-64-7 |
Výsledky specifikačního testu
Appearance | White to off-white to light yellow Solid or crystalline solid |
Infrared spectrum | Conforms |
Melting point | 58 to 64 °C |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...