Potassium ethyltrifluoroborate 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
444880010
444880050
ACRO444880010EA
3330
CZK
ACRO444880010
ACRO444880050
Potassium ethyltrifluoroborate 97%
Potassium ethyltrifluoroborate
Vzorec:
C₂H₅BF₃K Bod tání: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112713 CAS číslo: 44248-07-9 |
Výsledky specifikačního testu
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Melting point | ≥260 °C |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...