Tisk…

(S)-2-Aminopropanamide hydrochloride 95%

Dodavatel: Thermo Scientific

Synonyms: H-Ala-NH2.HCl, (S)-2-Aminopropionamide Hydrochloride, L-Alanamine Hydrochloride, H-Ala-NH₂·HCl, L-Alaninamide hydrochloride

442950010 442950050
ACRO442950010EA 1240 CZK
ACRO442950010 ACRO442950050
(S)-2-Aminopropanamide hydrochloride 95%
(S)-2-Aminopropanamide hydrochloride
Vzorec: C₃H₉ClN₂O
MW: 124,57 g/mol
Bod tání: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00066145
CAS číslo: 33208-99-0

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥94 to ≤106 %
Specific optical rotation +10° to +12° (20°C, 589 nm) (c=1 in MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR