Dimethyl-L-glutamate hydrochloride 99%, pure
Dodavatel: Thermo Scientific
Synonyms:
L-Glutamic Acid Dimethyl Ester Hydrochloride, H-Glu(OMe)-OMe.HCl
441940100
441940500
ACRO441940100EA
1670
CZK
ACRO441940100
ACRO441940500
Dimethyl-L-glutamate hydrochloride 99%, pure
Dimethyl-L-glutamate hydrochloride
Vzorec:
C₇H₁₃NO₄ MW: 211,64 g/mol Bod tání: 91…93 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038879 CAS číslo: 23150-65-4 |
Výsledky specifikačního testu
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Specific optical rotation | +25° to +27° (20°C, 589 nm) (c=5 in H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...