Potassium sulphite 90%, pure
Dodavatel: Thermo Fisher Scientific
440210010
440210025
440212500
ACRO440210010EA
4090
CZK
ACRO440210010
ACRO440210025
ACRO440212500
Potassium sulphite 90%, pure
Potassium sulphite
Vzorec:
K₂SO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00011387 CAS číslo: 10117-38-1 |
Výsledky specifikačního testu
Appearance | White Crystalline powder |
Titration Iodimetric | ≥88 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...