Tisk…

Benzyl N-(2-aminoethyl)carbamate hydrochloride 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: N-(2-Aminoethyl)carbamic acid benzyl ester hydrochloride, N-1-Z-1,2-diaminoethane hydrochloride, N-Z-Ethylenediamine hydrochloride, N-Carbobenzoxy-1,2-diaminoethane hydrochloride, N-Cbz-1,2-diaminoethane hydrochloride

439810010 439810050
ACRO439810010EA 1730 CZK
ACRO439810010 ACRO439810050
Benzyl N-(2-aminoethyl)carbamate hydrochloride 95%
Benzyl N-(2-aminoethyl)carbamate hydrochloride
Vzorec: C₁₀H₁₄N₂O₂
MW: 230,69 g/mol
Bod tání: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00270150
CAS číslo: 18807-71-1

Objednat

Výsledky specifikačního testu

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR