Tisk…

α-Ketoglutaric acid monosodium salt 98%

Dodavatel: Thermo Fisher Scientific
Varování

439350050 439350250 439351000
ACRO439350050EA 1730 CZK
ACRO439350050 ACRO439350250 ACRO439351000
α-Ketoglutaric acid monosodium salt 98%
α-Ketoglutaric acid monosodium salt
Vzorec: C₅H₅NaO₅
Storage Temperature: Ambient
MDL Number: MFCD00064196
CAS číslo: 22202-68-2

Objednat

Výsledky specifikačního testu

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1.0 %
UV at 430 nm A: ≤0.200
pH 3.6 to 4.0 (0.5 % in water)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR