2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt 99%
Dodavatel: Thermo Fisher Scientific
439030250
439031000
439035000
ACRO439030250EA
2280
CZK
ACRO439030250
ACRO439031000
ACRO439035000
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt 99%
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid sodium salt
Vzorec:
C₆H₁₄NNaO₅S Storage Temperature: Ambient |
MDL Number:
MFCD00067425 CAS číslo: 66992-27-6 |
Výsledky specifikačního testu
Appearance | White Powder |
Infrared spectrum | Conforms |
Elemental Analysis | 5.85 to 6.04 % (N) |
Titration with HCl | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...